Nalgene® Unitary™ Wash Bottles, Low-Density Polyethylene, Wide Mouth, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene® Unitary™
2402-0250 2402-0125 2402-0750 2402-1000 2402-0500
16651-573CS 7601.66 MXN
16651-573 16651-551 16651-595 16651-610 16651-609
Nalgene® Unitary™ Wash Bottles, Low-Density Polyethylene, Wide Mouth, Thermo Scientific
Frascos Frascos de lavado
Low-density polyethylene wash bottles enable efficient laboratory work.
Wide-mouth for faster, easier filling. No messy delivery tube. Contents can be dispensed without tipping or shaking. Dispensing tip can be cut back to increase flow. With polypropylene screw cap. Leakproof.
Precaución: Not recommended for long-term use with acetone.