VWR® Reclosable UV-Protectant Amber Bags
Proveedor: VWR
VWR®
MGUV3P0203 MGUV3P0305 MGUV3P0808 MGUV3P0608 MGUV3P0814 MGUV3P02509 MGUV3P0508 MGUV3P0406 MGUV3P1212
89005-326CS 2317.9 MXN
89005-326 89005-324 89005-334 89005-322 89005-332 89005-320 89005-330 89005-318 89005-328
VWR® Reclosable UV-Protectant Amber Bags
Bolsas Bolsas de almacenamiento
Ideal for storage and transport of light-sensitive products or medications.
  • Re-closable design
  • Amber tint protects against UV radiation
  • Easy to identify with amber tint
Re-closable 3 mil bags feature a wide, white zipper to allow for repeated use and provides protection against dust and moisture. Amber tint makes bags easily identifiable.
Certificaciones: Bags meet light transmission rates between the ranges of full shade at DIN 53775 and DIN 53387, as well as reduced shade at DIN 53375.