VWR® Wash Bottles
Proveedor: Avantor
VWR®
560070 560060 560050 560080
414004-227CS 3278 MXN
414004-227 414004-226 414004-225 414004-228
VWR® Wash Bottles
Frascos Frascos de lavado
LDPE wash bottle features a polypropylene screw cap and stem that are molded into a one-piece unit for leakproof operation.
  • LDPE construction
  • Provided with a polypropylene screw cap
  • Leakproof operation
Dispensing tube produces a fine stream, but can be cut for a coarser flow.