VWR® Autoclavable Biohazard Bags, Double Thick
Proveedor: VWR
VWR®
14220-088 14220-086 14220-084 14220-082
14220-082CS 4292.26 MXN
14220-082 14220-084 14220-086 14220-088
VWR® Autoclavable Biohazard Bags, Double Thick
Bolsas Bolsas de eliminación de residuos
Made of high-strength, highly temperature-resistant 3 mm polypropylene plastic that is autoclavable to 135 °C (275 °F).
  • Temperature-resistant polypropylene
  • Autoclavable
  • Biohazard warning label
Heavy-duty bags offer extra puncture resistance and high impact strength. Clearly labeled with proper biohazard warnings. Bright red color for quick identification.
Información para pedidos: Bags are supplied with 20.3 cm (8") twist ties.