VWR® Autoclavable Bags, 1.5 mil
Proveedor: VWR
VWR®
14220-022 14220-020 14220-016 14220-008 14220-006 14220-018 14220-002 14220-004 14220-010 14220-012 14220-014 14220-000 14220-024
14220-008CS 9038.34 MXN
14220-008 14220-012 14220-022 14220-000 14220-014 14220-002 14220-024 14220-016 14220-004 14220-018 14220-006 14220-010 14220-020
VWR® Autoclavable Bags, 1.5 mil
Bolsas Bolsas de eliminación de residuos
Autoclave bags provide an efficient, safe solution for the messy and potentially dangerous process of disposing of biohazardous waste material.
  • Bags are available clear or red with proper biohazard warnings, or clear without label
  • Rated for use at 135 °C (275 °F)
High-strength, highly temperature-resistant polypropylene construction allows perfect adaptability to standard laboratory disposal routines, including steam autoclaving.
Información para pedidos: Twist ties included.