Celstir® Suspension Culture Flasks, Wheaton
Proveedor: DWK Life Sciences
Celstir®
356890 356879 356884 356873 356882 356889 356875 356887 356876
62401-916CS 34094.43 MXN
62401-908 62401-916 62401-906 62401-914 62401-904 62401-912 62401-902 62401-910 62401-900
Celstir® Suspension Culture Flasks, Wheaton
Matraces Matraces centrífugas
For quality laboratory production and trypsinizing suspended cell cultures
Flask design provides an optimum inert cell growth environment. An adjustable blade impeller maximizes incubator space and provides maximum turbulence. A bottom dimple (on 125mL and larger sizes) prevents crushing of cells beneath the magnetic impeller and improves cell yield. Units larger than 250mL have 45mm sidearms, which can be used as air vents, media inlet/outlet ports, inoculation port, or to accommodate a pH probe. The screw cap assembly provides a positive seal using a fluoropolymer resin liner backed with silicone rubber. Easy to clean and assemble, the assembly can be autoclaved as a unit.
Designed for use with the Wheaton Biostir® Stirrers (see 12000-906 series).
Información para pedidos: Supplied with glass flask, fluoropolymer resin and glass impeller assembly, and fluoropolymer resin-lined top cap.