Nalgene® Heavy-Duty Funnels, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
4260-0040 4260-0060 4260-0100
30250-084CS 11288.14 MXN
30250-109 30250-062 30250-084
Nalgene® Heavy-Duty Funnels, Thermo Scientific
Embudos Embudos de filtración Embudos con filtro
Funnels are completely molded, unbreakable and with vent-ribs for even flow
The largest size is molded of high-density linear polyethylene (HDPE). Other sizes are low density polyethylene (LDPE).