VWR® Powder Funnels, Polypropylene
Proveedor: VWR, part of Avantor
VWR®
650040 650030 650010 650020
414004-269CS 376.94 MXN
414004-269 414004-272 414004-271 414004-270
VWR® Powder Funnels, Polypropylene
Embudos Embudos para polvos
Polypropylene funnels feature a 60° body angle and 58° internal rib angle for rapid filtration.
  • Autoclavable
  • Resistant to most chemicals
  • Polypropylene construction
External ribbing prevents air lock. Resistant to most chemicals, except strong oxidizing agents.