VWR® Laboratory Narrow Mouth Bottles, PP
Proveedor: VWR
VWR®
414004-182 414004-178 414004-177 582150 414004-179 414004-183 582140 414004-181 414004-180 582100 582090 582120 582130 414004-176 582080
414004-183CS 4031.04 MXN
414004-179 414004-178 414004-177 414004-176 89221-638 89221-632 89221-644 89221-634 89221-640 89221-630 414004-183 89221-642 414004-182 414004-181 414004-180
VWR® Laboratory Narrow Mouth Bottles, PP
Frascos Botellas de plástico
Specimen bottles are for general laboratory use, especially where sterilization is necessary.
  • Clear polypropylene construction
  • Screw enclosure included
  • Excellent chemical resistance
  • Can be stored in freezer to −40 °C (−40 °F)
  • Can be autoclaved with or without contents
Bottles are translucent and provide better clarity than polyethylene bottles
Note: Before autoclaving, set closure on top of container without engaging threads.
Información de suministro: Polypropylene screw closure included.