Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
4280-0550 4280-1500 4280-1100 4280-0900 4280-0700
30305-109CS 5523.12 MXN
30305-109 30305-040 30305-084 30305-062 30305-120
Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
Embudos Embudos de filtración Embudos Büchner
Sturdy two-piece construction
Separable for easier cleaning. Highly resistant to most chemicals. Autoclavable.