VWR® Autoclavable PE Bags, Double Thick
Proveedor: VWR
VWR®
14232-184 14233-764
14232-184CS 14737.8 MXN
14232-184 14233-764
VWR® Autoclavable PE Bags, Double Thick
Bolsas Bolsas de eliminación de residuos
Heavy-duty bags with extra puncture resistance and high impact strength.
  • Excellent puncture resistance
  • Clear sides allow for easy observation
High-molecular, high-density 4 mm polyethylene plastic. Autoclave up to 121 °C (250 °F). Ideal for sterilizing animal cages. Includes 20.3 cm (8") twist ties. All bags are clear. Meets many state and local regulations.