Nalgene® Separatory Funnels, Teflon® FEP, Squibb, Pear-Shape, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
4301-1000 4301-2000 4301-0500 4301-0125 4301-0250
30356-507CS 23283.69 MXN
30356-507 30356-529 30356-553 30356-551 30356-540
Nalgene® Separatory Funnels, Teflon® FEP, Squibb, Pear-Shape, Thermo Scientific
Embudos Embudos de separación
Funnels are non-stick, break-resistant, and can be used in radiological and hazardous separations
Compatible with common lab reagents and solvents. Impervious to practically any chemical used in a separatory funnel. Autoclavable except for stopcock assembly, which can be chemically disinfected.