Nalgene® PassPort™ Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene® PassPort™
2199-0004 2199-0008 2199-0032 2199-0016
16129-754CS 9716.78 MXN
16129-754 16129-756 16129-750 16129-752
Nalgene® PassPort™ Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Frascos Botellas de plástico
Safe, durable, cost-efficient bottles for packaging and transporting hazardous materials.
  • Molded into bottom: IP2 marking, material, volume, and registration notch
  • Heavy-duty walls
  • Leakproof
Evaluated at 15 psi (103 kPa) per 49 CFR 173.27 (c) (2), ICAO Technical Instructions Part 3-1; 1.1.6.1, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Recommended for customers who are designing, assembling, and certifying their own combination packaging. Split- and puncture-resistant. Wide mouth allows for easy filling of dry materials or liquids.
Precaución: Warning: Do not use under pressure or vacuum. The application of pressure or vacuum to products not designed for such use may result in product failure, damage to property, or personal injury.