PYREX® Buchner Funnels with Fritted Disc, Corning
Proveedor: Corning
PYREX®
36060-600C 36060-600F 36060-3LC 36060-2C 36060-60C 36060-3LF 36060-15F 36060-150F 36060-60M 36060-600M 36060-2LF 36060-150M 36060-30C 36060-350M 36060-15C 36060-60F 36060-15M 36060-2LC 36060-2F 36060-30F 36060-2LM 36060-30M 36060-350F 36060-350C 36060-3LM 36060-150C
89091-504CS 9500 MXN
89091-504 89091-502 30297-148 30297-149 89091-162 89091-160 89091-286 89091-166 89091-320 89091-342 89091-284 89091-164 89091-340 30296-201 89091-500 89091-324 89091-344 89091-168 89091-322 30297-150 30295-209 30296-210 89091-496 89091-298 89091-158 89091-498
PYREX® Buchner Funnels with Fritted Disc, Corning
Embudos Embudos de filtración Embudos Büchner
PYREX®, borosilicate glass, with sintered glass disc.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
Especially useful where filter paper would be attacked by material being filtered. Top edge is beaded.
Precaución: Disc should not be subjected to pressure differential in excess of 1.01 bar (1 atm).