VWR® Wide Mouth Laboratory Bottles, HDPE
Proveedor: VWR
VWR®
414004-113 414004-112 414004-115 414004-110 414004-114 414004-111
414004-115CS 2545.07 MXN
414004-115 414004-114 414004-113 414004-112 414004-111 414004-110
VWR® Wide Mouth Laboratory Bottles, HDPE
Frascos Botellas de plástico
Bottles hold liquid or dry material for applications in lab or field and are suitable for shipping liquids.
  • Excellent chemical resistance
  • Leakproof
  • Easy-to-pour wide mouth design
Can be stored in freezer to −100 °C (−148 °F). Excellent chemical resistance to alcohols, alkalis, and both diluted and concentrated acids. Thick walls afford durability. Shrink ring at neck ensures smooth interior so that contents pour fluidly, while sealing ring and semi-buttressed threads in closure ensure leak prevention. Bottles are translucent and produced via the injection blow molding process for exact features and bottle stability.
Información para pedidos: Polypropylene caps included.