VWR® Wide Mouth Laboratory Bottles, Amber HDPE
Proveedor: VWR
VWR®
581340 581320 581330 581310 414004-118 414004-121 414004-120 414004-117 581300 414004-116 414004-119 581350
414004-117CS 2884.54 MXN
414004-117 414004-116 89221-676 89221-678 414004-121 89221-672 414004-120 89221-674 89221-680 89221-670 414004-119 414004-118
VWR® Wide Mouth Laboratory Bottles, Amber HDPE
Frascos Botellas de plástico
Bottles are for general laboratory use, but are especially good for light-sensitive materials or photo- sensitive samples.
  • Ideal for light-sensitive materials or photosensitive samples
  • Leakproof
  • Easy-to-pour wide mouth design
Can be stored in freezer to −100 °C (−148 °F), and have excellent chemical resistance to alcohols, akalis, and both diluted and concetrated acids. Thick walls afford durability. Shrink ring at neck ensures smooth interior so that contents pour fluidly, while sealing ring and semi-buttressed threads in closure ensure leak prevention. Bottles are produced via the injection blow molding process for exact features and bottle stability.
Información para pedidos: Amber polypropylene caps included.