VWR® Laboratory Wide Mouth Bottles, PP
Proveedor: VWR
VWR®
582250 414004-126 582230 582210 582240 582220 414004-124 414004-123 414004-122 582200 414004-125 414004-127
414004-127CS 5500 MXN
414004-127 414004-126 414004-125 414004-124 89221-688 414004-123 414004-122 89221-684 89221-686 89221-690 89221-692 89221-682
VWR® Laboratory Wide Mouth Bottles, PP
Frascos Botellas de plástico
Specimen bottles are for general laboratory use, especially where sterilization is necessary.
  • Ideal for specimens and chemicals
  • Can be used as animal water bottles
  • Autoclavable
  • Leakproof
  • Easy-to-pour wide mouth design
Can be stored in freezer to −40 °C (−40 °F), and have excellent chemical resistance to alcohols, alkalis, and both diluted and concentrated acids. Thick walls afford durability, yet bottles are still squeezable. Shrink ring at neck ensures smooth interior so that contents pour fluidly, while sealing ring and semi-buttressed threads in closure ensure leak prevention. Bottles are translucent and produced via the injection blow molding process for exact features and bottle stability.
Información para pedidos: Polypropylene caps included.