VWR® Large Wide Mouth Bottles , PP
Proveedor: VWR
VWR®
414004-187 414004-188
414004-188CS 3902.98 MXN
414004-188 414004-187
VWR® Large Wide Mouth Bottles , PP
Frascos Botellas de plástico
Bottles are specifically designed for applications that require autoclaving of large volumes.
Note: Before autoclaving, set closure on top of bottle without engaging threads.
Información para pedidos: Bottle 414004-188 is supplied with a handle.
Información de suministro: 100-415 white polypropylene screw cap included.
Precaución: Bottles are not leakproof and are not recommended for shipping liquids.