VWR® Autoclavable PP Bags
Proveedor: VWR
VWR®
1102-0810 1102-2436 1102-1224 1102-0815 1102-1012 1102-0812 1102-1010 1102-2430 1102-1015
95042-563PK 1640.98 MXN
95042-552 95042-563 95042-550 95042-548 95042-558 95042-568 95042-556 95042-554 95042-564
VWR® Autoclavable PP Bags
Bolsas Bolsas de eliminación de residuos
Transparent 2 mm thick polypropylene bags are suitable for autoclaving hazardous and nonhazardous materials and waste.
  • Can withstand autoclaving temperatures up to 135 °C (275 °F)
Twist ties (included) allow proper closure for bag expansion and venting during sterilization.
Certificaciones: Meet strength requirements in ASTM D 1709-04 for 165g dart drop test.