VWR® Absorbent Pads
Proveedor: Avantor
VWR®
3803242VWR 3803240VWR
95057-860CS 7652.72 MXN
95057-860 95057-862
VWR® Absorbent Pads
Materiales para contención de derrames
Designed to soak up liquids, oils, and common solvents, abosorbent pads provide an excellent alternative to paper towels and gauze.
  • Strong, highly-absorbent, and lint-free
  • Efficient alternative to paper towels and gauze
Pads exhibit a cloth-like softness with no loose threads or fibers, making them ideal for use in coverslipping, slide preparation, and general laboratory cleanup. Available in two sizes, with the larger format individually folded for convenience.