Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
2110-0006 2110-0008 2110-0016 2110-0002 2110-0032
16120-953CS 8549.96 MXN
16120-956 16120-953 16120-962 16120-950 16120-959
Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Frascos Frascos Roller
Square, polypropylene wide mouth bottles are designed to replace glass serum and general-purpose storage bottles.
  • Resistant to acids, alcohols, and bases
  • Contact-clear bottles provide visible liquid level
  • Wide mouth for easy cleaning, sample filling, and retrieval
  • Square shape saves space and offers large labeling surface
  • Leakproof and repeatedly autoclavable
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Información para pedidos: Supplied with polypropylene screw caps assembled on bottles.