Corning® Storage Bottles, Square, Polycarbonate, Sterile, Corning
Proveedor: Corning
Corning®
431432 431430 431431 431433
89089-722CS 5086.57 MXN
89089-722 89089-756 89089-762 89089-816
Corning® Storage Bottles, Square, Polycarbonate, Sterile, Corning
Frascos Frascos de medios
PC, square.
  • More break-resistant than other glass or plastic bottles
  • Screened white enamel graduations are easier to see than moulded graduations
  • Large white marking spot for easier identification
  • Bottles can be autoclaved once at 121 °C and 15 psi (repeated autoclaving weakens PC and is not recommended)
  • Can be stored at –80 °C
These storage bottles are easier to handle, require less space on the shelf or in the autoclave, and are ideal for mixing, sampling and storage.