VWR® Easy-Use™ 9-Volt Accessory Adapter
4138
61161-384EA 1409.26 MXN
61161-384
VWR® Easy-Use™ 9-Volt Accessory Adapter
Easy-Use™ 9-Volt Accessory Adapter features a robust design and delivers accurate monitor readings.
  • 115 VAC
Adapter replaces the 9-volt battery and permits constant monitoring.