VWR® Moisture Meter
Proveedor: VWR
VWR®
4310
21800-020EA 12083.28 MXN
21800-020
VWR® Moisture Meter
Analizadores Analizadores de humedad
This moisture meter is designed to display the moisture content of wood, wood products, and paper.
  • Serial RS-232 Output
  • Traceable®
The memory contains nine material groups including fiberboard, construction board, and calibrations for 150 species of wood
Data logger captures and stores up to 8000 readings from all meters. Readings may be taken at intervals from 1 second to 99 hours. Stored readings may be downloaded to any PC and viewed. Data files can also be read on spreadsheets, databases, and statistical programs. Supplied complete with 91.4 cm (36") serial cable with D9F computer plug, 8.9 cm (31/2") diskette, and four AA alkaline batteries.
Data acquisition system captures and stores readings on any PC. Information can be imported into databases. Supplied with 8.9 cm (31/2") diskette and 91.4 cm (36") serial cable with D9F plug.
The unit provides for automatic as well as manual temperature compensations. It features a serial RS-232 output, which allows it to be connected to a computer or data logger for monitoring and keeping results. At the touch of a button, the instrument recalls highest, lowest, and average readings. A data HOLD button freezes the display to capture readings.
Probe dimensions: 18L x 2.5dia. cm (7 x 1")
Certificaciones: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Información para pedidos: Moisture meter is complete with probe, 10 spare pins, 9V alkaline battery, and serial computer output.