KIMAX® Funnels, Long Stem, 60° Angle, Kimble Chase, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMAX®
28980-150 28980-200 28980-250
30214-149EA 1813.27 MXN
30214-149 30214-160 89001-414
KIMAX® Funnels, Long Stem, 60° Angle, Kimble Chase, DWK Life Sciences
Embudos Embudos de filtración Embudos con filtro
Constructed with heavy uniform walls and strong, fire polished rims and stems for a long service life
These funnels are made of low extractable borosilicate glasses.
Funnels have a high resistance to chemical attack, mechanical shock, and thermal shock.
Certificaciones: Designed from ASTM Specification E1095, Type I, Class B requirements.