VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
Proveedor: VWR, part of Avantor
VWR® Tri-Pour®
25384-150 25384-154 25384-156 25384-158 25384-152 25384-160
25384-150CS 432.31 MXN
25384-150 25384-160 25384-154 25384-152 25384-158 25384-156
VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
Vasos de precipitados
Transparent polypropylene beaker with three pouring lips for dripless dispensing.
  • Polypropylene construction
  • Transparent
  • Graduated markings
  • Disposable design
Graduated, with accuracy of ±5%. May also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents. Write-on cardboard caps ensure spillproof, leak-resistant closure.