EP Scientific Depyrogenated Vials, Sterile, Thermo Scientific
Proveedor: Thermo Scientific
ST20-20 ST50-20 ST5-20 ST10-20 ST100-20 ST30-20 ST5-13 ST2-13
EPST50-20CS 27120 MXN
EPST20-20 EPST50-20 EPST5-20 EPST10-20 EPST100-20 EPST30-20 EPST5-13 EPST2-13
EP Scientific Depyrogenated Vials, Sterile, Thermo Scientific
Frascos Botellas de vidrio
Type I borosilicate vials are suitable for a number of uses where aseptic protocol is required.
  • Vials are assembled with butyl stoppers and aluminum seals
Certificaciones: Sterilization is performed in a FDA licensed facility.
Información para pedidos: Supplied with Certificate of Sterility and Pyrogen Test.