SP Wilmad-LabGlass Precision Bore Class A Burettes with Straight Stopcock, SP Industries
Proveedor: SP Industries
LG-3950-100
14203-424EA 7753.31 MXN
14203-424
SP Wilmad-LabGlass Precision Bore Class A Burettes with Straight Stopcock, SP Industries
Buretas
These borosilicate glass precision bore Class A burets feature a durable colored scale and have straight stopcocks with Standard Taper 2mm bore
Comply with accuracy requirement ASTM Specification E-287.