VWR® Pressure/Vacuum Gauge
Proveedor: VWR
VWR®
90027-60 90027-61
23609-220EA 18751.32 MXN
23609-222 23609-220
VWR® Pressure/Vacuum Gauge
Bombas Indicadores de vacío
Gauge measures in pounds per square inch, bar, meters of water, atmospheres, millimeters of mercury, inches of mercury, inches of water, and kilograms per square centimeter.
  • Traceable®
  • Measures in eight units
  • Stores up to 1000 readings
LCD display features 9.5mm (3/8") high digits
Data acquisition system captures, displays, and stores readings on any PC. Information can be imported into databases/statistical programs. Supplied with 31/2" diskette and 91.4 cm (36") serial cable with D9F plug.
Data logger captures up to 1000 readings from all meters. Six keys permit a range of different reading intervals. Stored readings can be downloaded to any PC. Supplied with cables, software, and four AA alkaline batteries.
Memory key recalls highest and lowest readings. Data hold button freezes the display to capture readings. RS-232 output allows connection to a computer or data logger for monitoring and storing results. Transducer sensor (included) has 6.4mm (1/4") NPT male-threaded stainless steel end. May be used with all gases and liquids compatible with 316 stainless steel. Gauge is accurate to 1% full scale plus one digit.
Certificaciones: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Información para pedidos: Gauge is supplied with 101.6 cm (40") cable, serial computer output, transducer sensor, and 9V battery.