PYREX® Gooch Crucibles, High Form, with Fritted Disc, Corning
Proveedor: Corning
PYREX®
32940-30F 32940-30C 32940-50M 32940-50F 32940-30M 32940-50C
89091-448CS 1300 MXN
89091-448 89091-328 89091-446 89091-330 23864-065 89091-450
PYREX® Gooch Crucibles, High Form, with Fritted Disc, Corning
Crisoles
Particularly adapted to analytical work in cases where precipitates are dried to constant weight at 110 °C (230 °F).
  • Made of PYREX® borosilicate glass
Also suitable for high temperatures, in which case it is advisable to heat in an electric furnace. In order to avoid strain, crucible should not be removed from furnace until the temperature has dropped to 250 °C (482 °F). Limit of use, 500 °C (932 °F).