KIMBLE® KIMAX® Gooch Crucibles, Low Form, with KIMFLOW® Fritted Disc, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMAX®
28260-303 28260-302 28260-152 28260-301
89001-366CS 769.35 MXN
89001-366 89001-370 89000-808 89001-368
KIMBLE® KIMAX® Gooch Crucibles, Low Form, with KIMFLOW® Fritted Disc, DWK Life Sciences
Crisoles
Especially suitable for precipitates to be dried to constant weight at 110°C
Note: Exposure to sudden temperature changes should be avoided.
For higher temperatures, an electric furnace is advised and crucibles should not be removed until temperature has dropped to 250°C.