SP Wilmad-LabGlass Gooch Crucibles, High Form, SP Industries
Proveedor: SP Industries
LG-5470-108 LG-5470-102 LG-5470-100
80089-456EA 1122.69 MXN
80089-456 80089-448 80089-450
SP Wilmad-LabGlass Gooch Crucibles, High Form, SP Industries
Crisoles
With fused-in fritted glass disc, these high form borosilicate glass Gooch crucibles are designed for analytical work involving precipitation, drying, and weighing.
They are particularly suitable when precipitates will not be dried at temperatures above 110°C, and may be used at higher temperatures. Fine (F), Medium (M), and Coarse (C) porosities available.
Precaución: Note: Avoid sudden temperature changes when heating or cooling.