VWR® Heavy-Duty Crucibles and Covers, Steel
Proveedor: VWR
VWR®
11648-192 11648-188 11648-191 11648-190 11648-189
82027-540CS 24978.03 MXN
82027-540 82027-538 82027-536 82027-542 82027-544
VWR® Heavy-Duty Crucibles and Covers, Steel
Crisoles
Low form crucibles and covers are designed for high-temperature applications, such as alkali and peroxide fusions.
  • Sheet steel construction
  • Loop handles for convenient handling
They are manufactured from sheet steel, and can withstand temperatures up to 500 °C (932 °F). Covers feature loop handles.
Información para pedidos: Crucibles and covers are sold separately.