VWR® Sample Bags for the Seward Stomacher® Blender
Proveedor: VWR
VWR®
EFT-1015A-VW SCR-7012-VW1 SCTO7012A-VW SCL41214-VW1 SCR-7012-VW1 SCLO7012-VW1 SCL-1520-VW SCL-4060-VW1 SCL47012-VW1 SCL47012-VW1 SCT-7012A-VW SCT-7012A-VW SCLO1520-VW SCL41214-VW1 SCLO1520-VW SCL41218-VW SCL-1520-VW SCLO7012-VW1 SCL41520-VW SCT-1520A-VW SCL-7012-VW1 EFT-1015A-VW SCL41520-VW SCTO7012A-VW SCL-4060-VW1 SCT-1520A-VW SCL-7012-VW1 EFT-7012A-VW EFT-6090A-VW
89085-562CS 8381.74 MXN
CA89085-567 CA89085-564 10048-888 CA89085-562 CA11216-892 CA11216-890 CA89003-820 CA11216-902 CA11216-900 10048-890 11216-898 76200-884 CA10048-880 CA10048-884 89085-566 CA10048-888 89085-562 89085-564 89003-820 11216-892 11216-890 11216-902 CA11216-898 11216-900 10048-882 CA10048-890 10048-880 10048-886 10048-884
VWR® Sample Bags for the Seward Stomacher® Blender
Bolsas Bolsas para homogeneizadores
Sample bags are designed to withstand the paddle action of Stomacher® lab blenders.
  • Pure or hybrid polyethylene construction
  • Highly transparent
  • Suitable for general-purpose blending or analysis testing
  • Some bags feature write-on strip
  • Malleable yet durable
FILTRA-BAG® bags are made of a mix of polyethylene and nylon.
SECURE-T® bags are made of heat extruded polyethylene tubing for internal sterility and to eliminate the need for side seals. Bags are malleable yet durable, with superior wall strength. May be used in general-purpose blending or analysis testing.
The LABRACK® storage rack, an ideal tool for storing samples, is designed to store up to ten 400 series blender bags, closed with a clip, in their upright position.
All standard and closure bags are made of highly transparent polypropylene and have wide-sealed bottoms to prevent leaks. Bags with 4 mm wall thickness are for analyses of firmer samples. Standard bags are made with heat-extruded virgin polyethylene tubing and feature a tear-off sterile barrier top to ensure sterility up to the time of use. Closure bags permit collection, transportation, blending, and storage of samples all within the same bag. They are available with regular wire or extra-strength flat wire closures. Some bags feature a write-on strip for easier sample identification.