VWR® Critical Print™ Cleanroom Pens and Markers
Proveedor: VWR
VWR® Critical Print™
BXPENBF WP1114-6BLU P1114-6RED P1114-6BLK
51280-876CS 3982.14 MXN
51128-048 51280-874 51280-876 51280-812
VWR® Critical Print™ Cleanroom Pens and Markers
Bolígrafos Bolígrafos para salas limpias
Manufactured with very low sodium to prevent sodium contamination.
  • Polyethylene barrels and caps
  • Nonsterile
Barrel and cap are made from high-density polyethylene.
Certificaciones: Suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms.