Marker II Superfrost® Marking Pens, Aspen Surgical
1450-16-PDC 1450-20-PDC
52878-121PK 888.27 MXN
52878-122 52878-121
Marker II Superfrost® Marking Pens, Aspen Surgical
Specifically designed to enable histology and cytology laboratories to permanently mark Superfrost slides, as well as ordinary frosted-end microscope slides and embedding cassettes
Write on glass, plastic, porcelain, metal, and paper. Resistant to most solvents and fixatives commonly used in histology/cytology procedures not requiring abrasive contact. Ink can be removed with various solvents if necessary. Markers should be tested with laboratory’s solvents and equipment before use on actual specimens.