Cleanroom Pens and Markers, Micronova
Proveedor: Micronova
PEN-11 PEN-20IRBB PEN-52IR PEN-01IR PEN-12 PEN-02IR PEN-51IR PEN-25IRX PEN-03 PEN-25IR PEN-01 PEN-02 PEN-20IR PEN-11IR PEN-12IR PEN-22IR PEN-03IR PEN-61IR PEN-25IR
68400-570CS 8521.5 MXN
76214-024 76214-299 76443-120 68400-693 76214-296 68400-572 68400-570 68400-691 76214-028 76214-026 76292-770 68400-568 68400-689 76203-464 76471-004 89107-766 89236-620 89107-767 89032-008
Cleanroom Pens and Markers, Micronova
Bolígrafos Bolígrafos para salas limpias
Micronova Cleanroom Pens feature low sodium, indelible ink and a special low particulate barrel to limit particle generation. Cleanroom Irradiated Sharpie® and Smear-Proof markers write permanently on most surfaces with quick drying ink that is fade- and water-resistant to further extend multiple project uses. Marks will not smear on reusable glass, metal, or porcelain lab-ware, but can be scrubbed off.
  • Indelible, low sodium ink pens
  • White, high-density polypropylene barrel
  • Holds up to IPA, bleach, and phenol wipe downs
  • Available Capped or Retractable
  • Choice of black, blue, or red ink
  • Available gamma irradiated for aseptic environments
Designed for documentation and labeling in cleanrooms and aseptic environments. Gamma irradiated pens and markers are suitable for Aseptic ISO 5 and other clean production areas. The unique polypropylene barrel is manufactured to limit particle generation and holds up to IPA, bleach, and phenol wipe downs. Available irradiated, capped, or click versions, with black, blue, or red ink.
Envase: Packed 10 pens per pack and 10 packs per case. Markers available in original capped and retractable styles are individually wrapped and double-bagged with four markers per pack and three packs per case.