PIG® Multi-Purpose Repair Putty, New Pig
Proveedor: New Pig
PIG®
PTY201
56620-098PK 6800 MXN
56620-098
PIG® Multi-Purpose Repair Putty, New Pig
Adhesivos
Made with the highest-quality epoxy resins to adhere to steel, glass, masonry and most plastics.
  • Sets in 20 minutes
  • Cures in 1 hour
  • Nontoxic
Convenient, premeasured epoxy has Parts A and B in one tube ready for kneading. Sets in 20 minutes and cures concrete-hard in 1 hour for drilling, tapping, sanding or painting. Nontoxic, VOC- and solvent-free putty can be used on potable water lines. 3- to 5-minute working time. A great addition to any toolbox or response kit.