VWR® Weighing Paper
Proveedor: Avantor
VWR®
20 60 5628 20 60 5626 20 60 5627
12578-201CS 26222.62 MXN
12578-201 12578-121 12578-165
VWR® Weighing Paper
Papeles de pesaje
Weighing paper is designed for analytical weighing with minimal influence on analytical results for weighing and transporting sample.
  • Clean and dust-free
  • Lightweight
  • Nitrogen-free
  • Smooth surface
Smooth surface is ideal for transferring weighed material. Clean, dust-free sheets. Paper is lightweight, yet strong enough to support analytical size samples.
Envase: All sizes of weighing papers are packaged 500 sheets per box.