Koolit® Gel Packs, Gel Pack Refrigerants, Cold Chain Technologies
Proveedor: Cold Chain Technologies
Koolit®
524-ST 564 616A 620E-SP 624 516 564 608A 524 612 532 512 624A 548 508A 648 632E 516SB4
89049-356CS 1164.79 MXN
89049-356 89049-364 75801-064 80080-904 89049-358 89049-362 89049-360 10813-684 33500-354 75784-520 33500-594 CA10813-684 33500-358 33500-356 89039-566 33500-350 33500-362 33500-360
Koolit® Gel Packs, Gel Pack Refrigerants, Cold Chain Technologies
Envases fríos
Formulated from nontoxic materials, these reusable Koolit® refrigerant gel packs maintain the thermal profile of temperature-sensitive products safely and more efficiently than ice.
The 500 Series gel packs contain a CMC-based gel formation, while the 500-ST Series packs contain a suppressed temperature gel formation. Both feature a puncture-resistant laminate foil film exterior. The 600 Series gel packs consist of a long-lasting polymer gel formulation in a 5mil thick white polyethylene pouch. They are ideal for one-time use or disposable applications.
Use the 0°C (32°F) gel formulation for the shipment of refrigerated products, and the –23°C (–10°F) formulation for products that need to stay below freezing temperatures.