PolarPack® Gel Packs, Ice Pack Refrigerant Gel Packs, Sonoco ThermoSafe
Proveedor: Sonoco
PolarPack®
PP8IN24BAGS PP48 PP16 PP24 PP8 PP6 PP42 TUFF24 PP12 PP3 PP32 PP20IN24BL
16000-068CS 2362.75 MXN
82029-734 89049-998 16000-068 10770-550 89495-998 89495-996 14100-476 89050-000 89024-674 75845-138 14100-478 89050-002
PolarPack® Gel Packs, Ice Pack Refrigerant Gel Packs, Sonoco ThermoSafe
Envases fríos
An economical alternative to wet ice packs, these nontoxic, reusable gel packs are enclosed in a durable 4.1 mil polyethylene film.
  • Reliable non-mechanical refrigeration
  • Ideal for transporting food and pharmaceuticals
  • Economical alternative to wet ice
PolarPack gel packs provide reliable non-mechanical refrigeration for the safe transport of food products, pharmaceuticals, and other medical products
The elongated design of PP8IN24BAGS allows for a protective wrap around sensitive specimen samples and diagnostic materials in vials and tubes during transport and shipping. Use in combination with an insulated shipper or envelope for additional assurance.
Refrigerant changes phase at 0 °C (32 °F) and returns to gel state upon thawing.