AC Medium Rounds, Clear, Wide Mouth, Graduated, Kimble®, DWK Life Sciences
Proveedor: DWK Life Sciences
5810858V-21 5810448C-21 5810238B 5810448B 5810448V-21 5810238C-26 5810858C-23 5810448C-23 5810858B 5810133C-24 5811670C-22 5810858C-21 5811670C-23 5810858C-26 5811670V-21 5810238C-25 5811670C-26 5810448C-26 5810238V-26 5811670V-26 5810238V-21 5810448C-22 5810133C-26 5810133V-21 5810238C-24 5810858V-26 5810133C-21 5810238C-21 5810858C-22 5811670C-21 5810133V-26 5811670B 5810448V-26 5810133C-25
89046-476CS 1254.14 MXN
89046-570 89046-472 89046-572 89046-474 89046-530 89046-574 89046-476 89046-510 89046-576 89046-478 89046-578 89046-556 89046-558 89046-508 89046-490 89046-480 89046-482 89046-582 89046-560 89046-484 89046-584 89046-562 89046-486 89046-586 89046-542 89046-564 89046-488 89046-544 89046-566 89046-588 89046-568 89046-526 89046-528 89046-506
AC Medium Rounds, Clear, Wide Mouth, Graduated, Kimble®, DWK Life Sciences
Frascos Botellas de vidrio
Versatile storage bottles provide an ideal containment for solid, liquid, or powdered specimens and samples.
The clear Type III soda-lime glass construction allows for constant sample viewing. Bottles feature graduated markings in both milliliters and ounces, and a wide mouth design for efficient sample addition and removal. Bottles are supplied with screw-on caps, unless otherwise noted.
Información para pedidos: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Corrugated cartons consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are packaged in bulk packs.