PolarPack® Gel Packs, Moisture Guard Refrigerant Gel Packs, Sonoco ThermoSafe
Proveedor: Sonoco
PolarPack®
MG48 MG12 MG16 MG24 MG32 MG8
89049-940CS 996.07 MXN
89049-994 89049-992 89049-942 89049-940 89049-996 89049-944
PolarPack® Gel Packs, Moisture Guard Refrigerant Gel Packs, Sonoco ThermoSafe
Envases fríos
An alternative to wet ice packs, these nontoxic, reusable gel packs have a multi-layered, cloth-like exterior to minimize and absorb surface condensation and protect temperature-sensitive products from moisture damage during shipping.
  • Multi-layered exterior
  • Absorbs surface condensation
  • Protects from moisture damage
Refrigerant changes phase at 0 °C (32 °F) and returns to gel state upon thawing.