VWR® 24- and 48-Place Storage Systems
Proveedor: Avantor
VWR®
3035-230-000
24270-230CS 5058.54 MXN
24270-230
VWR® 24- and 48-Place Storage Systems
Refrigerantes Refrigeradores de sobremesa
Racks float in your water bath and can accommodate dry ice for use with frozen samples on the bench.
  • Stackable
  • Autoclavable
  • Freezable
Microcentrifuge tube or cryo-vial storage system for 1.7 mL and 2.0 mL sizes. Autoclavable and freezable. These 24- and 48-place storage systems are made of polypropylene.