Eppendorf® PCR-Cooler
Proveedor: Eppendorf
022510509 022510525 022510541 022510525 022510509
62111-758EA 2795.05 MXN
62111-758 62111-760 CA62111-760 62111-762 CA62111-762
Eppendorf® PCR-Cooler
Refrigerantes Refrigeradores de sobremesa
More convenient and reliable than an ice bath, PCR-Cooler is ideal for preparing PCR reactions
Maintains a sample temperature of 0°C (32°F) for over one hour, if first pre-cooled at –20°C (–4°F) for two hours. Individual 0.2mL wells will change color when their temperature exceeds 7°C (45°F). Space-saving PCR-Cooler is easy to handle, and reduces the risk of contamination. Cone contour prevents samples from freezing. Cooler can accommodate individual tubes, tubes in a rack, tube strips, or a 96-well plate. Ideal for safely stopping reactions, and storing, transporting, and protecting sensitive samples. Available in violet changing to pink and dark blue changing to light blue.