Nalgene® Biohazardous Waste Containers, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
6370-0015 6370-0005 6370-0004
16211-134EA 4306.37 MXN
16211-136 16211-138 16211-134
Nalgene® Biohazardous Waste Containers, Thermo Scientific
Recipientes Recipientes para residuos
Autoclavable polypropylene waste containers are ideal for secondary containment of biohazardous waste materials.
  • Convenient one-step sterilization, with no need to remove autoclavable bag
  • Port opening in cover allows convenient one-handed disposal of materials, limiting exposure to infectious waste
  • Holds standard-size autoclavable bags
  • Red universal biohazard symbol is molded in English and Spanish
  • Leakproof side and bottom construction
Containers comply with OSHA 29CFR 1910.1030 for use as protection against bloodborne pathogens.