VWR® Biohazard Incinerator Cartons
Proveedor: VWR
VWR®
13205-1002 13205-1001
56617-810PK 866.99 MXN
56617-810 56617-807
VWR® Biohazard Incinerator Cartons
Cajas Cajas para la eliminación de residuos
A pair of tabs along the top edge of this cardboard receptacle keep the clear 0.038 mm (1.5 mil) thick polyethylene biohazard bag from slipping out of place.
  • Allows safe, convenient disposal and incineration of biohazardous waste
  • Negates the need to transfer contents
  • The snug-fitting lid has a flap for easily inserting waste material
Printed with large, bright, biohazard graphics and instructions in four languages (English, Spanish, French, German), it is packaged flat to save space and shipping costs. The carton and inner bag can be assembled in seconds.
Optional, reusable carton cover (for floor model only) provides a wider opening 20.3 x 24.1 cm (8 x 9.5") permitting safer and more sanitary deposit of wastes in to the container. Opens quickly and easily by pressing the built-in release tab. No batteries are required. Features a smooth 10.1 x 10.1 cm (4 x 4") area on top for applying labels. Autoclavable lid is color-coded for easy identification.