VWR® Broken Glass Disposal Cartons
Proveedor: VWR
VWR®
13204-6001 24653-1002 24653-1001
89088-054CS 5309.67 MXN
89088-054 56617-801 56617-804
VWR® Broken Glass Disposal Cartons
Cajas Cajas para la eliminación de residuos
Disposal cartons provide a safe, convenient place to throw away broken glassware and residual liquids.
  • Easy to assemble
  • Disposable cardboard construction
  • Safe and convenient disposal of broken glassware
  • Color-coded lids
The entire unit can be disposed of as a whole once the safety cap is pulled into place. The autoclavable lid is color-coded for easy identification, and the cartons are available in floor and benchtop models. Optional reusable carton covers, for the floor model only, provide a wider opening of 20.3×24.1 cm (8×9.5") for safer and more sanitary deposit of wastes into the container. The included polypropylene bag is 2 mil thick.
Pressing the built-in release tab opens the carton. No batteries are required. The carton features a smooth area on top for applying labels.
Información para pedidos: A carton cover for the floor model glass disposal carton must be purchased separately.
Envase: Cartons are packed flat to save space.