Durable Insulated Containers, Less than 5 cu. ft., Sonoco ThermoSafe
Proveedor: Sonoco
ThermoSafe®
301 300 304 302 450 390 303 422
35742-104EA 12905.04 MXN
35742-101 89496-030 35742-104 73320-006 35742-417 89496-028 16191-311 35742-100
Durable Insulated Containers, Less than 5 cu. ft., Sonoco ThermoSafe
Refrigerantes Arcón hielo seco
ThermoSafe Storage Chests provide mobile, non-electrical, low temperature maintenance for a wide variety of laboratory and field applications.
  • Deluxe containers provide years of trouble-free service
  • Rugged polyethylene interior and exterior casing with super-efficient, non-deteriorating urethane foam insulation
  • Rounded interior corners for easy clean-up
  • Spring-loaded handles fold down when not in use
These dry ice chests are ideal for demanding cryogenic and laboratory testing, as well as the transport and storage of temperature sensitive specimens.