Multi-Use One-Piece Sharps Collectors, BD Medical
Proveedor: BD Medical
305577 305610 305665 305601 305490 305488 305615 305489 305616 305491 305609 305666 305602 305477
BD305488CS 5199.97 MXN
BD305577 BD305610 BD305665 BD305489 BD305488 BD305477 BD305490 BD305491 BD305609 BD305601 BD305666 BD305602 BD305616 BD305615
Multi-Use One-Piece Sharps Collectors, BD Medical
Recipientes Contenedores de objetos punzantes
One-step disposal systems feature puncture-resistant polypropylene containers to prevent needle penetration
Units feature handles, tight-fitting vented lids, and safety necks so sharps cannot pop back. Translucent tops allow easy viewing of contents. Containers have sterile-indicator tape on bottom to indicate sterilization completion.
For accessories, see BD305451 series.
One-way valve guards against accidental spilling. Containers may be autoclaved and compacted, or incinerated.